Trouble Shooter Aerosol Cleaner
Fournisseur: 3M
CHINTENS
MMMACHINTENSEA 272 EUR InStock
MMMACHINTENS
Trouble Shooter Aerosol Cleaner
Produits nettoyants Détergents et Nettoyants de Surfaces
Trouble shooter cleaners are cleaning sprays for removing tough stains from difficult-to-reach areas. They easily remove heavy soil, wax, or floor finish build-ups from hard surfaces.
  • Easy-to-apply with the directional nozzle
  • Easily removes stains from water resistant surfaces
  • Clings to vertical surfaces without dripping
  • Designed for use with the 3M™ Doodlebug Cleaning System for best results
Remove tough stains from difficult-to-reach areas with 3M™ Trouble shooter cleaners. The non-ammoniated formula penetrates and loosens stains quickly, meaning no lengthy downtime and no rinsing or neutralising is needed. Thick white foam clings to vertical surfaces and resists dripping, simply rinse it off with clean warm water. For use on a variety of water-resistant surfaces including ceramic, quarry tile, marble, fibreglass, and plastics, making them ideal for cleaning skirtings, floor edges and corners, uneven floors, stairwells, and showers.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité