Scotch-Weld™ DP804 EPX Super Clear Acrylic Adhesive
Fournisseur: 3M
DP804
MMMADP804EA 544 EUR InStock
MMMADP804
Scotch-Weld™ DP804 EPX Super Clear Acrylic Adhesive
Adhésifs
The Scotch-Weld™ EPX™ Bonding system is a discharging system for mixing two-component mixed adhesives. The main agent and the curing agent with the cartridge are mixed and discharged by a special hand gun and a special nozzle.
  • Easy mixing and application with the help of the EPX gun
  • 3 minutes of working time/4 minutes of handling time
  • Exposure at temperatures from 0 to 100 °C
  • Easy to apply
Stable adhesion can be obtained by eliminating the hassle of troublesome six-liquid mixing.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité