Scotch-Weld™ EPX Pneumatic Applicator
Fournisseur: 3M
EPXG50P
MMMAEPXG50PEA 453 EUR InStock
MMMAEPXG50P
Scotch-Weld™ EPX Pneumatic Applicator
Distributeurs d'adhésifs liquides et pistons
Quickly and easily metre, mix and apply two part adhesives with Scotch-Weld™ EPX pneumatic applicator.
  • Pistol grip provides comfortable handling and precise control
  • Measure, mix and apply
This applicator is designed to efficiently apply 3M™ Scotch-Weld™ Structural Adhesive Duo-Pak Cartridges for fast and simple applications using a pneumatic connection. The pneumatically charged system is ideal for large volume dispensing. It’s an effective system for bonding, joining, gluing, attaching, assembling, encapsulating, potting and sealing applications.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité