Fit Test Solution
Fournisseur: 3M
FT-32
MMMAFT-32EA 300 EUR InStock
MMMAFT-32
Fit Test Solution
Respirators
Replacement fit test solution for use with 3M™ qualitative fit test apparatus kits FT-10 (Sweet) and FT-30 (Bitter). The choice between the bitter and sweet test kit is a personal one, however a small percentage of the population are unable to detect saccharin, in which case the bitter test must be used.
  • Fit test solution; bitter taste
  • 55 ml bottle
  • For occupational use only
Test kits and accessories designed to help organisations meet regulatory fit testing requirements for tight fitting respirators. This product can be used to validate face to respirator seal on any disposable respirator or half mask respirator.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité