Scotch-Brite™ 350 Extra-Thick Delicate Surface Cleaning Pads
Fournisseur: 3M
HP350
MMMAHP350EA 103 EUR InStock
MMMAHP350
Scotch-Brite™ 350 Extra-Thick Delicate Surface Cleaning Pads
Lingettes Lingettes nettoyantes
Scotch-Brite™ Extra-thick delicate surface cleaning pads 350 are made from a high-quality synthetic fibre with a non-woven open structure. Non-abrasive cleaning particles are spread uniformly throughout the thickness of the fibre and linked with a durable, long-lasting resin.
  • Delicate and non-aggressive on surfaces
  • Flexible and adaptable
  • Long-lasting
  • Easy to rinse
Clean delicate surfaces without leaving scratches. These extra-thick cleaning pads are flexible and adaptable for delicate and non-aggressive cleaning on surfaces.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité