Rapid readout biological indicator for EO 1294, Attest™
Fournisseur: 3M MEDICAL
M1294E
MMMEM1294EEA 352 EUR InStock
MMMEM1294E
Rapid readout biological indicator for EO 1294, Attest™
Indicateurs de stérilisation
Four-hour, green cap, Rapid Readout Biological Indicator for EO Sterilization . Biological indicators and test packs that monitor the effectiveness of the EO sterilisation process and provide results within four hours.
  • After sterilisation, the vial is 'crushed' to join the growth media with the processed spore strip
  • The BI is incubated for 4 hours for a fluorescent readout
  • Test packs present a challenge to the sterilization process as do the AAMI recommended packs
  • Every load monitoring and quarantining can eliminate recalls
The self-contained BI for all EO cycles consists of a Bacillus atrophaeus spore strip; sealed glass ampule with growth medium and bromothymol blue pH indicator dye system; green colour-coded cap with holes for sterilant penetration and a spunbonded olefin (polyethylene) filter as a bacterial barrier, and a chemical indicator on the label that changes to green when processed.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité