Hygiene monitoring and management system, Clean-Trace™
Fournisseur: 3M Food Safety
LM1
MMMPLM1EA 3232 EUR InStock
MMMPLM1
Hygiene monitoring and management system, Clean-Trace™
Systèmes de surveillance Tests de contrôle de l'hygiène
3M™ Clean-Trace™ Luminometer is a simple, quick and reliable hand held diagnostic tool to verify cleaning effectiveness and capture, store and manage data for later analysis.
  • Re-engineered for improved precision and accuracy
  • Easy to use and fast time to result
  • Robust for the manufacturing environment
  • System includes the 3M™ Clean-Trace™ Luminometer and 3M™ Clean-Trace™ Hygiene Monitoring and Management Software to help you monitor testing results over time
  • Ergonomic design for simple one-handed operation
  • Wireless data transfer
  • 23 languages for multilingual
More Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité