Molecular detection assay 2 - Cronobacter
Fournisseur: 3M Food Safety
MDA2CRO96
MMMPMDA2CRO96EA 885 EUR InStock
MMMPMDA2CRO96
Molecular detection assay 2 - Cronobacter
Systèmes de surveillance Tests de contrôle de l'hygiène
3M™ Molecular Detection Assay 2 - Cronobacter is used with the 3M™ Molecular Detection System for the rapid and specific detection of Cronobacter species in enriched powdered infant formula, raw materials to manufacture powdered infant formula, infant cereals, and environmental samples.
  • Accurate, next-day results for the detection of Cronobacter spp in powdered infant formula and related samples
  • One assay protocol and fewer steps than most tests reduces repetitive tasks and makes it easy to train employees
  • Ready-to-use and pre-dispensed reagents provide convenience and minimise the chance of human error
  • Color-coded assay tubes by organism and matching software screens allow for better tracking and easy identification
  • The ability to run all assays simultaneously and the flexibility to test up to 96 samples in each run helps you operate more efficiently
  • Real-time amplification and detection process identifies positive results in as early as 15 minutes and allows you to make critical decisions faster
As with all test methods, the source, formulation and quality of enrichment medium can influence the results. 3M has evaluated the 3M Molecular Detection Assay 2 Cronobacter with Buffered Peptone Water ISO.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité