Swab-sampler with 10 ml neutralising buffer
Fournisseur: 3M Food Safety
RS96010NB
MMMPRS96010NBEA 114 EUR InStock
MMMPRS96010NB
Swab-sampler with 10 ml neutralising buffer
Écouvillons Écouvillons d'échantillonnage
3M™ Swab-sampler with 10 ml neutralising buffer is a convenient and quick collection device for environmental sampling and testing over small surface areas.
  • Neutralising buffer filled vial is ready to use for environmental sampling
  • Swab reaches into corners and hard-to-reach areas for increased accuracy
  • Convenient design helps improve sampling consistency from tech-to-tech and plant-to-plant
  • Bottles have ample headspace for sample addition and vortexing
  • Self-contained swab and vial for increased productivity and efficiency
Sample collection is easyMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité