Smart® FFP Masks with Comfortable Nose Seal
Fournisseur: Moldex
238501
MOLD238501EA 77.5 EUR InStock
MOLD238501
Smart® FFP Masks with Comfortable Nose Seal
Respirators Masques à usage unique Demi-masques fitrants contre les particules
Smart FFP masks are designed with a soft foam nose cushion for a comfortable fit. A soft, skin-friendly inner fleece layer and a stretchy woven strap provide additional comfort for the wearer.
  • Reliable protection against particles
  • Soft nose cushion for enhanced wearing comfort
  • ActivForm® 3D design for a comfortable and secure fit
The flexible ActivForm® 3D design adapts automatically to the shape of the wearers face and their facial movements. As a result, our Smart masks are easy to wear and sit securely and comfortably on the skin at all times, without the need for a nose clip. Our DuraMesh® structure preserves the shape of the mask under even the most testing conditions.
Smart FFP masks offer reliable protection against particles.
Shelf life from date of production: 5 years.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité