Digital microscope, Panthera DL
Fournisseur: MOTIC
1100403300061
MOTI1100403300061EA 3350 EUR InStock
MOTI1100403300061
Digital microscope, Panthera DL
Microscopes
This upright digital microscope features an integrated 4 MP WiFi camera that enables users to view and capture their images on an independent tablet using the free MotiConnect App.
  • Widefield high eyepoint eyepieces with diopter adjustment on both eyepieces and rubber eyecups
  • Reversed coded quintuple nosepiece with Motic LightTracer (Intelligent Light Management)
  • CCIS® Plan achromatic objectives UC PL (4×, 10×, 40× S, 100× S-Oil)
  • Coaxial coarse and fine focusing system with tension adjustment
  • Compact mechanical stage with built-in low position rackless coaxial stage control and sample holder
  • Köhler illumination with LED (3 W) and halogen (6 V/30 W) interchangeable illuminator
Motic LightTracer: A combination of a coded nosepiece and a digital light intensity knob for a digital illumination control. It memorises light intensity for each objective so that users don’t have to adjust it again.
Live display mode / Wi-Fi Live display – 1920×1080 pixels (Full HD)
Live display mode Ethernet Data transfer – 1920×1080 pixels (Full HD)
Information de livraison: Supplied ready-to-use with eyepieces, objectives, power cord, blue filter, immersion oil (5 ml), allen hexagonal key, vinyl dust cover, HAL bulb and LED module, Motic Images Plus 3.0 software for Windows, OSX and Linux, MotiConnect App for iOS and Android, and calibration slide.
Commander maintenant
COMMANDE
SPÉCIFICATIONS
ACCESSOIRES
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité