USB Camera, Moticam® S20
Fournisseur: MOTIC
1100600101401
MOTI1100600101401EA 1410 EUR InStock
MOTI1100600101401
USB Camera, Moticam® S20
Caméras
High Grade sCMOS USB Camera Moticam S20 for microscopy for an ultimate imaging result.
  • Fast frame rates
  • sCMOS sensors back-illuminated sensor technology
  • USB 3.1 interface
  • Motic Images Plus 3.0 software
Maximum details - If it is about maximum resolution for display or printing, if resolution in display or for printing is in your mind, Moticam S20 is your solution.
Informations de commande: To be purchased separately - C-mount adapter not included to be purchased from the same brand of your microscope, compatible with your microscope model and fitting with the size of the Chip of your Moticam.
Information de livraison: Includes CS ring adaptor, USB 3.1 cable, Motic 4-dot calibration slide, Motic Images Plus 3.0 for Windows/OSX/Linux.
Attention: Minimum computer requirements – 2 GHz dual-core, RAM memory 2 GB, video memory min. 512 MB.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité