Respirator, miniSCAPE
Fournisseur: M S A
10038560
MSAA10038560EA 92.3 EUR InStock
MSAA10038560
Respirator, miniSCAPE
Respirators Masques réutilisables Demi-masques
Using cutting-edge technology, the miniSCAPE respirator provides minutes of safety when seconds count. The portable respirator uses the new-generation TabTec filter, fits in a user's pocket and protects against numerous gases and vapors.
  • Convenient, portable respirator small enough to fit in a user's pocket
  • For search and rescue purposes
Unit uses the TabTec filter to provide 5 minutes of protection and is housed in a rugged carrying case.
The miniSCAPE is fitted with a comfortable silicon mouthpiece directly connected to the filter housing. A nose clip is attached to the unit by a securing cord.
The miniSCAPE is stored in an impact resistant plastic case, which can be quickly and easily opened by pulling the sealed tab located on the front.
Certifications: DIN 58647-T7 (ABEK-5) duration: 5 minutes; CA 13.182.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité