Safety spectacles, PERSPECTA 9000
Fournisseur: M S A
10045517
MSAA10045517EA 8.8 EUR InStock
MSAA10045517
Safety spectacles, PERSPECTA 9000
Brillen
Incredibly light, resilient and durable protective eyewear for all-day wear. Snug-fitting wrap-around lens provides perfect impact protection around the eyes.
  • Special nose-pads secure glasses to the face, eliminating the problem of slippage while providing a pillow-soft fit
  • Tight-fitting unitary front with dual 9.75 base curvature offers an unobstructed field of vision
  • Revolutionary hard and soft material co-injected temple for improved comfort
Weight: 26 g
Certifications: EN 166
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité