Disinfectant and virucide, Virkon® S
Fournisseur: MARKET SOURCE PART PROCESS
100-125-5KG
MSPP100-125-5KGEA 71.93 EUR InStock
MSPP100-125-5KG
Disinfectant and virucide, Virkon® S
Produits nettoyants Désinfectants
Virkon® S is a science-based biocidal technology for animal health disinfection, which has been specifically engineered to provide an effective, flexible and convenient, modern formulated disinfectant solution for commercial livestock producers, veterinary hospitals and farmers.
  • Broad spectrum
  • Effective against 61 strains of virus, 33 strains of bacteria and seven strains of fungi
It is versatile and can be used for non-porous hard surfaces, agricultural production equipment, vehicles, wheels, boot disinfection and day-to-day farm disinfection.
It is recognised worldwide as the leading disinfectant of choice for the emergency disease control of Avian Influenza (bird flu). It is independently proven to be effective against lethal strains of the highly pathogenic Avian Influenza virus.
Avian Influenza (fowl plague), also referred to as bird flu, is a potentially devastating disease, predominantly of chickens and turkeys, although the virus can also affect game birds (pheasants, partridge and quail), ratites (ostrich and emu), psittacine and passerine birds.
Attention: Use biocides safely. Always read the label and product information before use.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité