ADVANTAGE 24 WELL GOLD ANODIZED ALUMINUM PHOTOREDOX BLOCK ASSEMBLY, INCLUDES VIAL RACK, BOTTOM & TOP COVERS, PFA SHEET, RUBBER MATS AND SCREWS FOR 8X30MM VIALS 1 * 1 Pce
Fournisseur: MARKET SOURCE PART PROCESS
ADVANTAGE 24 WELL GOLD ANODIZED ALUMINUM PHOTOREDOX BLOCK ASSEMBLY, INCLUDES VIAL RACK, BOTTOM & TOP COVERS, PFA SHEET, RUBBER MATS AND SCREWS FOR 8X30MM VIALS 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
ADVANTAGE 24 WELL GOLD ANODIZED ALUMINUM PHOTOREDOX BLOCK ASSEMBLY, INCLUDES VIAL RACK, BOTTOM & TOP COVERS, PFA SHEET, RUBBER MATS AND SCREWS FOR 8X30MM VIALS 1 * 1 Pce MSPP24253 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou