Wide Storage Tower
Fournisseur: MULTIROIR
104461
MULC104461EA 177 EUR InStock
MULC104461
Wide Storage Tower
Mobilier Armoires de stockage
This wide three-drawer polypropylene tower is ideal for storing various materials, whether they are consumables or stationery.
  • 3 transparent drawers, each with a 30 L capacity
  • 4 self-assembly multidirectional wheels
  • Lid reinforced with a metal bar, capable of supporting up to 100 kg
  • Drawers with large handles
  • Anti-drop stops on drawers
Made entirely of polypropylene, this storage tower allows for the storage of various products. Additionally, it is resistant to temperature variations, chemicals, and impacts. Therefore, it can be placed in both laboratories and offices without any issues. Lightweight, this storage tower gains mobility once its four wheels are installed.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité