Folding Backrest Trolley
Fournisseur: MULTIROIR
65021
MULC65021EA 373 EUR InStock
MULC65021
Folding Backrest Trolley
Chariots
Trolley with fold-down backrest to help transport bins, boxes and cartons.
  • Tray in reinforced PP, 35 mm thick
  • 100 mm dia. silent castors
  • Two fixed castors + two swivel castors with brakes
  • 300 kg load capacity
  • Black
  • Ideal for 600×400 Europe containers
  • Delivered unassembled for easy assembly
This folding backrest trolley is made of polypropylene. It allows for the movement of a load not exceeding 300 kg. It is equipped with four wheels (two fixed and two multidirectional). This trolley is delivered unassembled. A few minutes are required for assembly.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité