Garder ma session ouverte ?
Expiré dans 
La session est expirée
Votre session a expiré. Pour votre sécurité, VWR vous a déconnecté.
Voulez-vous vous connecter à nouveau?

Muffle furnaces with lift up door, Tmax 1100 °C and controller C550, LT 3/11 - LT 60/11

Fournisseur: Nabertherm
Nabertherm
455-0965EA 4080 EUR InStock
455-0965 455-1778 455-1679 455-2506 455-1682 455-1267 455-1268 455-1061 455-1480 455-1260 455-1680 455-1768 455-0956 455-0955 455-1769 455-0957 455-1672 455-1770 455-1058 455-1674 455-1673 455-1478 455-1775 455-1258 455-1477 455-1059 455-0963 455-0962 455-1259 455-1776 455-1270 455-1052 455-1470 455-1051 455-1472 455-1053 455-1471
Muffle furnaces with lift up door, Tmax 1100 °C and controller C550, LT 3/11 - LT 60/11
Étuves Fours
The LT 3/11 - LT 60/11 muffle furnaces are the ideal choice for daily laboratory use. These high quality models have an advanced and attractive design, and are very reliable. The muffle furnaces have a lift up door as standard.

  • Textured stainless steel housing
  • Dual shell housing for low external temperatures and high stability
  • Lift up door (LT) with hot surface facing away from the operator
  • Exhaust air outlet on the rear wall of furnace
  • Solid state relays provide low-noise operation
  • Defined application within the constraints of the operating instructions
  • NTLog Basic for Nabertherm controller: Recording of process data with USB-flash drive

Heating is from two sides by means of ceramic heating plates (heating from three sides for muffle furnaces LT 24/11 - LT 60/11). These ceramic heating plates with an integral heating element are protected from fumes and splashing, and are easy to replace. Only fibre materials which are not classified as carcinogenic according to TRGS 905, class 1 or 2 are used.

The furnace is equipped with C550 professional controller with touch screen display, featuring 10 freely adjustable programs and 20 segments and two extra functions. These controllers are PID microprocessor controlled with a self-diagnostic system and integrated process documentation. The C550 controllers can automatically activate the vent with fan or catalyst (not model LT 15).

Informations de commande: All units require a 230 V single-phase power supply, except LT 24, LT 40 and LT 60 (400 V 3/N/PE). Optional accessories e.g. a vent, vent with fan or catalytic converter, over-temperature limit controller with adjustable temperature cut out for thermal protection Class 2, in accordance with EN 60519-2 as temperature limiter to protect the oven and load, protective gas connection on the rear wall of furnace. Manual or automatic gas supply systems are also available. Corresponding models with fold down doors, different Tmax and other controllers are also available. Please search the website for more details.

Attention: Maximum continuous operating temperature should be 50 °C below the 'Max. temp.' of the model.
Note heat up times: If connected at 230 V 1/N/PE rsp. 400 V 3/N/PE.
Outer dimensions include opened lift up door.
* Temperature uniformity of ± 5K in the empty work space for temperatures >800 °C.

Commander maintenant
 

COMMANDE

Documentation

Saisissez les détails du certificat

Saisissez le numéro de lot :

Choisissez parmi les numéros de lot/lot récents :

Ou
Examinez et comparez les solutions de rechange
VWR a trouvé des produits alternatifs qui peuvent vous sauver jusqu’à . Par unité d'article.  Pour comparer les détails d'un produit, sélectionnez jusqu'à 3 alternatives ci-dessous et cliquez sur Comparer les produits sélectionnés. Pour ajouter des articles à votre panier, entrez une quantité et cliquez sur Ajouter au panier.

Produit original :

Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité