Nalgene® Measuring Cylinders, Variety Pack
Fournisseur: Thermo Fisher Scientific
3662-1234
NALG3662-1234EA 224 EUR InStock
NALG3662-1234
Nalgene® Measuring Cylinders, Variety Pack
Éprouvettes Éprouvettes graduées
Measuring cylinders, made of PP. A convenient and cost effective way to get started setting up a new lab, assembling student lab kits, or refreshing an existing lab.
  • Large blue octagonal base prevents rolling and tipping
  • Molded-in graduations are silk-screened for easy reading
  • Excellent chemical resistance for a wide variety of solutions
  • Impact resistant at temperatures above −40 °C
  • Autoclavable, graduated
All sizes have a generous pour spout and molded-in graduations that are inked for easy reading. No meniscus allows accurate readings every time.More Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité