Nalgene® Measuring Cylinders, Variety Pack
Fournisseur: Thermo Fisher Scientific
3662-1234
NALG3662-1234EA 224 EUR InStock
NALG3662-1234
Nalgene® Measuring Cylinders, Variety Pack
Éprouvettes Measuring Cylinders
Measuring cylinders, made of PP. A convenient and cost effective way to get started setting up a new lab, assembling student lab kits, or refreshing an existing lab.
  • Large blue octagonal base prevents rolling and tipping
  • Molded-in graduations are silk-screened for easy reading
  • Excellent chemical resistance for a wide variety of solutions
  • Impact resistant at temperatures above −40 °C
  • Autoclavable, graduated
All sizes have a generous pour spout and molded-in graduations that are inked for easy reading. No meniscus allows accurate readings every time. Won’t break or chip like glass labware. Suitable for food and beverage use.
Certifications: ASTM, CFR 21 Part 177.1520, E1272.
Informations de commande: Includes seven cylinders, one of each size: 10, 25, 50, 100, 250, 500 and 1000 ml.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité