Blender bags, Whirl-Pak®
Fournisseur: NASCO FORT ATKINSON
B01239WA B01421WA B01474WA
NASCB01239WA NASCB01421WA NASCB01474WA
Blender bags, Whirl-Pak®
Sachets Sachets d'homogénéisation
Blended, virgin polyethylene film provides exceptional clarity, and superior strength for use in homogenizer. HACCP, EPA, USDA, FDA approved for food contact.
  • Plus 1 sterilisation, post-manufacturing sterilisation: Increased sample integrity, perforated closure to maintain sterility
  • Continuous perimeter seal: Maximum protection against leaks
This bag is specially designed for use with homogenizer blenders. It is made from extra-heavy polyethylene and has special features to make sample testing easier and more efficient.
Shelf life from sterilisation date found on the sterility documentation: 5 years from sterilised date found on your sterility confirmation.
Certifications: Approved by USDA, FDA, EPA and HACCP. Food safe.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité