Tube rack, neoRack®
Fournisseur: NEOLAB
240121634
NEOL240121634EA 0 EUR InStock
NEOL240121634
Tube rack, neoRack®
Portoirs Tube Racks
This tube rack is open at the front and are pierced by three frame levels or square grids, which are inserted and latched into the side and back sections.
  • Alphanumeric identification of the openings
  • Stackable
  • Colour coded
  • Can be quickly assembled and disassembled
  • Can easily be stowed in a cupboard or drawer
  • Plugged together blue/red
Made of durable polypropylene, temperature resistant and autoclavable from 90 to +120 °C.
The sides and back as well as the bottom floor are blue and the two upper levels to fix the tubes are colour coded.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité