Drain cover, PIG® Essentials
Fournisseur: NEW PIG
DC7676
NEWPDC7676EA 293 EUR InStock
NEWPDC7676
Drain cover, PIG® Essentials
Matériaux absorbants
Drain covers deploy quickly and create a tight, temporary seal over drains to keep out unwanted liquids.
  • Durable, tear-resistant top layer withstands repeated use and abrasion
  • Flexible, tacky bottom layer tightly seals drain openings
  • Chemical-resistant polyurethane resists water, oil, and many chemicals
  • Flexible cover moulds to uneven surfaces to create an instant, temporary drain seal that prevents pollution
High-visibility red top colour draws attention to spills for increased safety, while a dark bottom colour hides dirt and grime. Unlike messy alternative clay mats, it is easy to clean and reuse.
For best performance, make sure draincover overlaps drain opening at least 8 cm on each edge (max. drain size is 30×30 cm).
Attention: Temperature limit: use from –18 to 71 °C (0 to 160 °F) up to 12 months.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité