Sani-Trax™ disinfectant mat
Fournisseur: NEW PIG
FLME175
NEWPFLME175EA 41.8 EUR InStock
NEWPFLME175
Sani-Trax™ disinfectant mat
Tapis de sol Tapis de sol fixes
Clean and sanitise shoes and prevent contamination from entering controlled areas with this disinfecting footbath.
  • Thousands of flexible fingers dislodge contaminants from footwear using automatic scrubbing action
  • As traffic passes, rubber tips bend under pressure to immerse soles in disinfectant solution, keeping contamination from reaching food processing areas
  • Thick surrounding border stands 6.5mm higher than finger-scrapers to contain up to 4,5 litres of disinfectant solution
  • Convenient mat size that fits most doorways features suction cups on underside to prevent mat from shifting
  • Made from a tough natural rubber compound designed to resist to most chemicals and variations in temperature
Use at doorways, packaging areas, laboratories, locker rooms, break rooms, refuse areas and receiving rooms - anyplace where spread of contamination must be limited.
Attention: Use quaternary ammonium, iodine, chlorine or any other sanitiser but always test its concentration. Most sanitisers are concentrated and must be diluted before use in a footbath. Follow label directions for appropriate dilutions.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité