PIG® CHAT MAT® Roll
Fournisseur: NEW PIG
MAT619-DEC
NEWPMAT619-DECEA 134 EUR InStock
NEWPMAT619-DEC
PIG® CHAT MAT® Roll
Matériaux absorbants
Keep floors dry whilst delivering a message that can't be ignored.
  • Clearly visible message is repeated down the length of the roll to help create a safer environment for workers
  • Durable top layer prevents wear, even under heavy foot traffic
  • Under the Chat Mat top layer is the proven eight-layer, 100% polypropylene construction of PIG Mat
  • Exclusive dimple pattern speeds wicking of liquid throughout mat for faster, easier cleanup
  • Absorbs and retains oils, coolants, solvents and water
  • Flame-resistant material won't burn immediately like cellulose mats; melts when exposed to high heat whilst on the job
  • Can be wrung out and incinerated after use to reduce waste or for fuels blending
Easy-tear perforations let you take only what you need, so you use less mat and save money. High-visibility yellow colour is easier to see in poorly lit areas to minimise tripping hazards; alerts workers to potential dangers and works in progress.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité