Granules absorbantes Crysta-lite
Fournisseur: NEW PIG
P45-1
NEWPP45-1EA 12.8 EUR InStock
NEWPP45-1
Granules absorbantes Crysta-lite
Matériaux absorbants
An economical, light weight, non-combustible loose absorbent for spillages of water, oils, organic solvents, and most acids. PIG® Crysta-lite Absorbent will absorb many times its own weight.
  • Can be used as an absorbent media in enclosed containers for the disposal of liquid toxic waste substances
  • Remains granular, even when saturated, to provide excellent retention; stands up to heavy traffic without releasing liquids
  • Material is quarried, crushed, processed and screened to create a particle size of 3.0mm to 6.0mm
  • The "expansion" of the product generates countless tiny air bubbles within the structure creating the ultra light weight of PIG® Crysta-lite Absorbent
  • High surface area and excellent, fast-absorption properties give Absorbent the ability to create a low-slip surface
  • Chemically inert product is ideal for the absorption and removal of chemical spills, oil and water spillages on floors, nuclear waste, hazardous chemicals and more
  • Non-toxic mineral product is safe to handle and use without any risk
Weight: 3,5 kg per bag
Volume bag: 30 L
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité