Utility tray with grate, PIG®
Fournisseur: NEW PIG
PAK123-BK
NEWPPAK123-BKEA 157 EUR InStock
NEWPPAK123-BK
Utility tray with grate, PIG®
Matériaux absorbants
Durable PE utility tray is ideal for temporary storage of leaking parts or cores awaiting degreasing, repair or pickup. It helps to save bench space as large and heavy parts like batteries, grimy cores waiting for rebuild and leaky containers can be stored safely on the floor.
  • Strong and durable, chemical resistant
  • Ribbed bottom prevents batteries and containers from resting in spilled liquids
  • Heavy-duty walls and ribbed bottom provide strength and prevent warping
  • Includes heavy-duty grate to elevate parts and containers out of deeper liquids in sump
Weight: 11,6 kg
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité