Absorbent granules, PIG® PEAT
Fournisseur: NEW PIG
PLP404
NEWPPLP404EA 36.7 EUR InStock
NEWPPLP404
Absorbent granules, PIG® PEAT
Matériaux absorbants
All-natural peat moss absorbent pulls in oil-based liquids but not water. The super-absorbent, non-leaching activated sphagnum peat moss absorbs up to 30-litres of oil per bag. It is made for in and outdoor use and even removes oily sheens on floors.
  • Incinerable to help reduce waste
  • Non-abrasive composition will not harm floors or expensive machinery
  • Silica-free composition does not pose the health hazards associated with breathing in clay dust
  • Lightweight absorbent is easier than clay to carry and use without injury
Attention: Not recommended for oil spills on water or for use with corrosive liquids.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité