PIG® Bio-fluids solidifying absorbent powder
Fournisseur: NEW PIG
PLP503
NEWPPLP503EA 248 EUR InStock
NEWPPLP503
PIG® Bio-fluids solidifying absorbent powder
Matériaux absorbants
Fast-acting absorbent captures biofluids like blood, urine and vomit and "locks" them inside for cleaner, safer disposal.
  • Permanently solidifies to lock in absorbed liquids so they won't leak or liquefy when the absorbents are handled or placed under pressure
  • Contains chlorine to kill pathogens and reduce odors
  • Clean scent helps reduce offensive odors during and after cleanup
  • Environmentally-friendly product absorbs biological fluids including blood, urine and vomit
  • Forms a solid, non-expanding mass for easier, less-expensive recovery and cleanup
  • Shaker bottle makes it easy to apply absorbent directly onto spill area
High Btu value produces less than 0.1% ash content, ideal for fuels blending and incineration.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité