PIG® FIRE-DRI absorbent granules
Fournisseur: NEW PIG
PLPE270
NEWPPLPE270EA 16.6 EUR InStock
NEWPPLPE270
PIG® FIRE-DRI absorbent granules
Matériaux absorbants
PIG® FIRE-DRI is a granular organic product, suitable for a wide range of industrial applications. With its fireproof property, it can be used for liquids and applications where the use of fire resistant products is desired.
  • PIG® FIRE-DRI is a vegetable absorbent made up of resin wood dust, dried at high temperatures and processed to develop a fireproof behaviour
  • After absorbing liquid spills, it leaves clean, dry and safe surfaces, free from oily sheen
  • This fire-retardant material is capable of absorbing more than 3kg of oil per 1kg of product
  • This natural, renewable resource absorbent has a low environmental impact since it can be eliminated by incineration
  • Woodfibre is PEFC certified, which shows commitment to forest sustainable development
  • Non-toxic material contains no silica and is safe to handle and use
Non-abrasive, light-weight material is ideal for petrol stations, garages, airports, service companies, cleaning companies, railway stations, transport companies, emergency equipment, firemen, army and more.
Weight: 6 kg per bag
Volume Bag: 30 L
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité