Leak diverter bucket kit with absorbents, PIG® Roof Leak Diverter
Fournisseur: NEW PIG
TLS667-TR
NEWPTLS667-TREA 256 EUR InStock
NEWPTLS667-TR
Leak diverter bucket kit with absorbents, PIG® Roof Leak Diverter
Matériaux absorbants
This leak diverter kit features a pail lid with a removable plug to allow the drainage hose to be inserted without having to remove the lid. This can be used as a collection container for diverted liquid.
  • Non-permeable tarp is vinyl coated for strength and durability
  • Resistance to punctures, UV and mildew for long life
  • Flow rate is 12,6 litres/minute when used with a 12 mm ID clear drainage hose
  • Adjustable straps allow fast setup and response
  • Perfect for rapid response to overhead leaks
Informations de commande: Includes a drainage hose (635 mm), 4× adjustable hanging straps (1524 mm), a bucket (246 litres), a 'CAUTION' absorbent mat pads (30 packs) and two all purpose wipers (25 packs).
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité