Circulators, Clifton, NE4 series
Fournisseur: NICKEL ELECTRO
NE4-56P
NICKNE4-56PEA 3530 EUR InStock
NICKNE4-56P
Circulators, Clifton, NE4 series
Thermostats à circulation Systèmes à circulation ouverte Bains avec circulateurs détachables
These circulating water baths are available in two ranges for circulating just water up to 99 °C or oil up to 130 °C. Operating temperatures below ambient can be achieved using an optional refrigerated dip cooler. Units have touch pad controls with easy wipe clean fronts which are chemical resistant and splash proof.
  • Powerful circulation pump, provides a constant flow
  • Convenient ‘push-fit and lock' fittings on both inlet and outlet ports for secure external water circulation
  • Control options, single point temperature control or process control with two temperatures and a time setting
  • Automatic setting alarms for protecting bath contents, audible and visual indication
  • Anti-bacterial paint finish that actively inhibits bacterial growth
Maximum pump capacity: 10 L/min, 147 mbar
*RT = Ambient
Informations de commande: Supplied with a stainless steel mounting bridge and removable stainless steel shelf, but without racks or lids (lids are recommended when working above 60 °C or below ambient). Optional dip cooler is required for operating at below ambient temperature. Please order required accessories separately.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité