Cleanroom face masks, BioClean DB™, PE
Fournisseur: Nitritex
BDBN-G
NITRBDBN-GEA 326 EUR InStock
NITRBDBN-G
Cleanroom face masks, BioClean DB™, PE
Masques Masques pour salles propres
The non-sterile pouch style mask has knitted headbands and all seams are ultrasonically bonded. The mask has inner and outer facing layers constructed of spunbond non woven polyester with the middle filter layer made from meltblown polyester.
  • Integral neck guard
  • Latex-free
  • Patented spacer maintains clear breathing space
  • Ultrasonically sealed edges
  • Low-linting
  • Fully enclosed malleable nose-band
Suitable for use in ISO class 4, cleanroom environments.
Store in a dry, cool place (<40 °C) away from direct sunlight and fluorescent light.
Certifications: Manufactured in a facility holding ISO 9001:2015. Processed in an NEBB certified ISO class 4 cleanroom environment. Sampling and inspection in accordance with ISO 2859-3:1992.
Conditionnement: 50 pieces per sealed inner PE bag; six inner bags per sealed outer PE bag; one outer bag per lined carton (300 pieces).
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité