RABS/isolator mittens, BioClean™ Sterile Nitrile
Fournisseur: Nitritex
GGL30NITM9_U GGL30NITM9
112-5409EA 402 EUR InStock
112-5409 NITRGGL30NITM9
RABS/isolator mittens, BioClean™ Sterile Nitrile
Gants
These sterile RABS and isolator mittens are manufactured from nitrile with incredibly low levels of particles and excellent ESD properties.
  • Tested against ASTM D6978-05 for handling chemo drugs
  • 100% air leak tested
  • Suitable for autoclaving
  • Gamma irradiated (Sterility Assurance Level (SAL) of 10⁻⁶)
  • Can be sanitised by VHP or IPA
  • Ultra-clean surface ensures product protection
  • Specially designed to minimise hand fatigue
Designed for use in product contact areas, the GGL mittens are fully validated for sterility with an SAL (Sterility Assurance Level) of 10⁻⁶.
Suitable for ISO Class 4 and EU GMP Grade A cleanrooms.
Certifications: EN 388:2016 (4101X), EN ISO 374-1 Type A (AJKNOT), EN ISO 374-5 VIRUS, EN 421; Cat. III
Food contact approved
Tested against permeation standard ASTM D6978-05 for cytotoxic drugs.
Conditionnement: Triple-bagged: One piece per sealed inner PE bag; one inner bag per sealed second inner PE bag; one second inner bag per sealed outer PE bag; 20 outer bags per lined inner white Correx polyethylene box (20 pieces).
Attention: No glove provides complete protection against all chemicals. Users must test the gloves against the particular chemicals and environment in which they will be used.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité