Sterile sleeve/glove system, BioClean™
Fournisseur: Nitritex
GSG10NITXLMA_U GSG10NITXLMA
112-6124EA 578 EUR InStock
112-6124 NITRGSG10NITXLMA
Sterile sleeve/glove system, BioClean™
Gants
Clean and sterile sleeve/glove system, nitrile sleeve attached to an ambidextrous polychloroprene (BioClean S-BFAP) glove by a channel ring and O-ring.
  • Sleeve and glove tested against ASTM D6978-05 for handling chemo drugs
  • Ultra-clean surface ensures product protection
  • Sleeve 100% inspection and air leak tested (prior to being guillotined)
  • Sleeve suitable for autoclaving
  • Gamma irradiated (Sterility Assurance Level (SAL) of 10⁻⁶)
  • Sleeve can be sanitised by VHP or IPA
  • Fits port size 8"/203 mm
Compatible with ISO Class 4 and EU GMP Grade A cleanrooms.
* Length of the complete system (glove + sleeve)
Certifications: EN ISO 374-1 Type C, EN ISO 374-5 VIRUS, EN 421; Cat. III; AQL 0,65
Food contact approved
Tested against permeation standard ASTM D6978-05 for cytotoxic drugs.
Conditionnement: One system - consisting of sleeve, glove, and channel ring/O-ring assembly packed in inner PE bag; two inner bags (two systems – one L and one R) packed per outer PE bag; 10 outer bags (20 systems) per lined white Correx box. Sold by the pair, individually packaged.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité