MEA cleanroom looped face mask, BioClean™
Fournisseur: Nitritex
MEA210-2
NITRMEA210-2EA 602 EUR InStock
NITRMEA210-2
MEA cleanroom looped face mask, BioClean™
Masques Masques pour salles propres
Non sterile ear loop face mask.
  • High bacterial, viral, and particle efficiency filtration
  • Fully enclosed malleable noseband
  • Ultrasonically sealed edges
  • Ear loops for quick and easy donning
  • Clip connector for secure fastening at back of head available to purchase separately
Providing high particle and bacterial filtration efficiency, the BioClean™ MEA Cleanroom Looped Face Mask is manufactured from cleanroom compatible materials to reduce contamination into the controlled environment. The mask features ear loops for quick donning and doffing. (Clip connector to allow for quick and secure fastening at back of head are available to purchase separately; see datasheet).
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité