Anti-IgA Mouse Monoclonal Antibody [clone: HISA43] (Janelia Fluor® 646)
Fournisseur: Novus Biologicals
NBP311510JF646
NOVUNBP311510JF646EA 1100 EUR InStock
NOVUNBP311510JF646
Anti-IgA Mouse Monoclonal Antibody [clone: HISA43] (Janelia Fluor® 646)
Antilichamen
This monoclonal antibody is specific to heavy chain of IgA and shows minimal cross-reaction with heavy chains of other immunoglobulins. It is reactive with both IgA1 and IgA2 subclasses of Alpha heavy chain. It reacts with the third constant domain (CH3) of the alpha chain of IgA molecules.
Immunoglobulins are four-chain, Y-shaped, monomeric structures comprised of two identical heavy chains and two identical light chains held together through inter-chain disulfide bonds. The chains form two domains, the Fab (antigen binding) fragment and the Fc (constant) fragment. Immunoglobulin A (IgA)More Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité