Crimp top vials, 8 mm
Fournisseur: THERMO NATIONAL SCIENTIFIC
C4008-632R
NSCAC4008-632REA 112 EUR InStock
NSCAC4008-632R
Crimp top vials, 8 mm
Fioles
These crimp/snap top low volume sample vials can be used with 6, 7 and 8 mm autosampler trays. They are made of borosilicate glass, type 1, class A.
  • Crimp top seal minimises exposure between sample solvent and septum
  • Without write-on patch
Reusable sleeves support the crimped, sealed vials in the correct position in autosampler carousels or in racks and allow movement of the vials as a unit to injection positions in both GC and HPLC autosamplers.
Certifications: These vials meet all requirements of US, EU and JPN Pharmacopia.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité