Waving shaker, analogue
Fournisseur: OHAUS
30391968
OHAU30391968EA 1100 EUR InStock
OHAU30391968
Waving shaker, analogue
Agitateurs Agitateurs rotatifs
This analogue waving shaker is an easy and economical option for all the waving needs in laboratory. Ideal for cell culture and blotting applications, and is designed to be used in a variety of environmental conditions. The aluminium base offers durability and added stability
  • Takes up less space and easily fits into most hoods and incubators
  • Microprocessor control provides tilt adjustment which allows user to easily adjust waving angle from 0 to 16° while unit is operating
  • Speed ramping feature slowly increases speed to desired set-point to avoid splashing
  • Timer will automatically stop rocking motion when timer reaches zero
  • Spill-resistant design ensures fluids are channelled away from internal components
  • Can be used at −10 to 60 °C (max. 80% relative humidity, non condensing) in cold rooms and incubators
Independent control knobs for speed, tilt, and time, allow for easy adjustments. Audible signal will activate when system detects an obstruction or tray overload.
Applications include staining and destaining gels, hybridisation procedures, hematology, and blotting techniques.
Information de livraison: Supplied with a 356×279 mm non-skid rubber mat and a detachable, 3-wire cord and plug.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité