Mini vortex mixers
Fournisseur: OHAUS
30392110 30392117 30392124 30392131
OHAU30392110EA 259 EUR InStock
OHAU30392110 OHAU30392117 OHAU30392124 OHAU30392131
Mini vortex mixers
Agitateurs Agitateurs Vortex
Ergonomic mini vortex mixers with features to suit different applications, these compact units provide optimum comfort and minimal stress.
  • Continuous or touch mode operation (except fixed speed unit which has touch mode only)
  • Can be used at 4 to 40 °C (from 20 to 85% relative humidity, non condensing)
  • Pulsing model has unique action which reduces heat generation and ensures efficient mixing and disruption
  • Digital and pulsing units have timer, 1 s to 160 h
The analogue unit features low speed start up for gentle shaking or high speed vigorous mixing.
The fixed speed unit starts mixing when the cup head is pressed down, it provides vigorous vortexing at full speed.
The digital model is ideal for applications that need reproducable results as the microprocessor control maintains the set speed. The timer displays elapsed time or can be programmed to a user-defined time limit and will switch off when elapsed time reaches zero. Two modes of operation; continuous mode when using accessory attachments or touch mode which activates mixing when depressing the cup head.
The pulsing model provides powerful pulsing, ensuring complete cell disruption within minutes for glass bead procedures. Capable of complete cell disruption of samples in just minutes. The pulsing action reduces heat generation while providing more effective mixing and disruption. Displayed time counts up during continuous operation and counts down during timed runs.
Information de livraison: All models supplied with a cup head, a 76 mm head with cover. Pulsing vortex mixer is supplied with a stainless steel microtube holder with built in cup head and holds up to 1,5 to 2 ml microtubes.
Commander maintenant
COMMANDE
SPÉCIFICATIONS
ACCESSOIRES
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité