Vortex mixer, digital
Fournisseur: OHAUS
30392136
OHAU30392136EA 1030 EUR InStock
OHAU30392136
Vortex mixer, digital
Agitateurs Agitateurs Vortex
This heavy-duty digital vortex mixer is ideal for applications that demand repeatable results. Mixer features touchpad controls and LED displays for accurate speed and time (minutes and seconds) results. The heavy-duty design and efficient motor allow this mixer to operate in continuous duty mode and handle all accessories over the entire speed range.
  • Variable speed microprocessor control provides consistent uniform mixing action
  • Timer will display elapsed time or, when programmed to user defined time limits, the unit will shut off when time reaches zero
  • Display will show last used settings, even after power has been turned off
  • Can be used at 4 to 40 °C (from 20 to max. 85% relative humidity, non condensing)
Two modes of operation: 'Touch' mode for mixing tubes when cup head or universal holder with cover is depressed, or 'On' mode when using any of the accessory attachments for continuous operation.
Information de livraison: Supplied a cup head, universal holder with cover, foam microtube insert for (38) 1,5 to 2,0 ml microtubes, and a detachable, 3-wire cord and plug.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité