Vortex mixer, analogue
Fournisseur: OHAUS
30392141
OHAU30392141EA 772 EUR InStock
OHAU30392141
Vortex mixer, analogue
Agitateurs Agitateurs Vortex
This heavy-duty analogue vortex mixer is a variable speed analogue mixer that is designed for continuous duty. The heavy-duty design and efficient motor allow this mixer to operate in continuous duty mode and handle all accessories over the entire speed range.
  • Economical alternative to digital models
  • Can be used at 4 to 40 °C (from 20 to max. 85% relative humidity, non condensing)
Two modes of operation: 'Touch' mode for mixing tubes when cup head or universal holder with cover is depressed, or 'On' mode when using any of the accessory attachments for continuous operation.
Information de livraison: Supplied with a cup head, universal holder with cover, foam microtube insert for (38) 1,5 to 2,0 ml microtubes, and a detachable, 3-wire cord and plug.
Commander maintenant
COMMANDE
SPÉCIFICATIONS
ACCESSOIRES
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité