Guardian 5000 hotplate
Fournisseur: OHAUS
30500533
OHAU30500533EA 468 EUR InStock
OHAU30500533
Guardian 5000 hotplate
Appareils chauffants Plaques chauffantes
Reliable hotplate with exceptional safety features for everyday use.
  • SafetyHeat™ system protects the laboratory
  • Easy to read LCD display with everything you need to know
  • Designed for durability and reliability with SmartHousing™
OHAUS Guardian 5000™ hotplates are designed for safety and simplicity in all research, academic and industrial applications. The design ensures proper temperature control of the top plate or your sample when using the optional temperature probe.
These heating models include a Single Point Calibration feature for multiple setpoints. Ideal for everyday heating applications.
Informations de commande: Supplied without optional temperature probe which must be ordered separately.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité