Endeavor™ 5000 Light Duty Digital Orbital Shakers
Fournisseur: OHAUS
30770915
OHAU30770915EA 1045 EUR InStock
OHAU30770915
Endeavor™ 5000 Light Duty Digital Orbital Shakers
Agitateurs Agitateurs orbitaux
Endeavor™ Light Duty Shakers are available in 3 models: 3 and 20 mm orbit, or dedicated microplate model. All models can be used in incubators and cold rooms. Stable, accurate, long-term shaking is ensured with the triple eccentric drive and maintenance-free brushless DC motor. Integrated overload and unbalanced detection to protect samples.
Applications: Microbiology, molecular biology, ELISA assays, cell cultures, immunoassays, protein studies, bacterial and yeast cultures, staining/destaining, blotting techniques, DNA studies, solubility studies, Environmental and Food Sciences, chemistry, general mixing.
Display: Bright, backlit LCD display for speed and time, timer will count up elapsed time or when set, timer will count down and shut off shaking when time reaches zero, green indicator light illuminates when unit is shaking, easy to navigate menu for advanced features.
Operation: Simple knob control for timer and shaking, tray with attachment points for flask clamps, tube racks and non-slip rubber mat included, optional accessories include universal platform and over 20 flask clamps and tube racks. One model dedicated for microplates.
Communication: RS232 interface provides two-way communication for data logging and unit control.
Construction: Durable chemical-resistant polymer housing, sturdy easy to clean aluminum standard tray with attachment points for accessories, IP 21 ingress protection, robust triple eccentric drive for stable shaking, brushless DC motor, control knobs, on and off switch.
Design features: Visual and audio alarm when timer counts down to zero or an error occurs, small footprint to save bench space, advanced microprocessor control slowly ramps the shaker tray up to the desired speed and avoids splashing, easy to attach optional accessory platform.
Commander maintenant
COMMANDE
SPÉCIFICATIONS
ACCESSOIRES
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité