MagBinder® Fit24 Nucleic Acid Purification System
Fournisseur: OMEGA BIO-TEK
B1-001-24
OMEGB1-001-24EA 20070 EUR InStock
OMEGB1-001-24
MagBinder® Fit24 Nucleic Acid Purification System
Systèmes de purification des acides nucléiques
Elevate your laboratory experience with the MagBinder® Fit24 Nucleic acid purification system. Efficiency, affordability, precision, and reliability, all in one powerful instrument.
  • Ease-of-use – Automation-ready cartridges pre-filled with reagents
  • Convenience – No reagent preparation and buffer dispensing
  • High-quality purification – Up to 4 ml samples lysed offline eluted in as little as 50 μl
Simple and efficient – Experience efficient nucleic acid purification with the MagBinder® Fit24 Nucleic acid purification system. This affordable, state-of-the-art instrument utilises 24 magnetic rods coordinated to pick-up, transfer, and release magnetic particles within reagent cartridge wells, ensuring a reliable and streamlined process for DNA and RNA purification. Accommodating 1 to 24 samples simultaneously, the MagBinder® Fit24 is compatible with a range of sample types.
Turnkey solution – Get to processing samples faster with seamless integration of specially designed MB Fit24™ cartridges. The MagBinder® Fit24 comes complete with validated, pre-loaded scripts for MB Fit24™ cartridges, allowing faster set-up time and less manual intervention.
Flexibility at its finest – Designed to work seamlessly with Omega Bio-tek Mag-Bind® kits, the MagBinder® Fit24 presents a flexible, programmable platform. The user-friendly interface empowers effortless programming on new protocols directly on the instrument.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité