E.Z.N.A.® Tissue DNA Kit, E.Z.N.A®
Fournisseur: OMEGA BIO-TEK
D3396-03
OMEGD3396-03EA 939 EUR InStock
OMEGD3396-03
E.Z.N.A.® Tissue DNA Kit, E.Z.N.A®
Réactifs pour acides nucléiques Nucleic Acid Purification Kits and Reagents
Isolate DNA from tissues, buccal swabs, cultured cells, whole blood, body fluids, paraffin-embedded tissues, and mouse tail snips using mini spin columns.
  • Rapid - DNA isolation in less than 20 minutes post-lysis
  • Versatile - Single kit for multiple sample types
  • Specialised buffer system - Optimised buffers for higher yields
  • Safe - No phenol/chloroform extractions
  • High-quality - DNA is suitable for a variety of downstream applications
The E.Z.N.A.® Tissue DNA Kit offers a versatile and cost-effective method for the isolation of DNA from a wide variety of samples including fresh or frozen animal cultured cells and tissues, buccal swabs, whole blood, mouse tail snips and more. The DNA purification process is simplified with HiBind® Mini Column technology into four quick 'lyse-bind-wash-elute' steps and can be accomplished in less than 20 minutes post-lysis.
This convenient spin-column format avoids time-consuming steps like alcohol precipitation, use of toxic compounds such as phenol and chloroform and allows for multiple samples to be processed in parallel. DNA purified using this kit is ready for most downstream applications such as PCR, sequencing, genotyping, southern blot analysis, and restriction enzyme digestion.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité