HBO Fluorescence Microscope, IM-300
Fournisseur: OPTIKA S.R.L
IM-300FL4
OPTKIM-300FL4EA 11766 EUR InStock
OPTKIM-300FL4
HBO Fluorescence Microscope, IM-300
Microscopes
Advanced inverted microscope for brightfield and fluorescence observations with Semi-Apo IOS LWD U-PLAN F objectives to enhance the visibility of the sample and increase the overall contrast.
The HBO fluorescence illuminator provides an outstanding flexibility of use, standing the blue and green filter sets (supplied as standard) for Auramine, FITC, GFP and YFP (with blue filter set) plus Rhodamine, Texas Red and TRITC (with the green one), yet giving the possibility to combine any other specific filter sets for future upgrade.
Transmitted light through the exclusive X-LED⁸ to ensure great-looking, rich and high-quality specimen view.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité