Heat inactivated and filtered animal serum containing a detergent and a preservative.Used in ELISA assays for diluting standards, reagents and for blocking non-specific interactions. 1 * 100 mL
Fournisseur: OriGene
Heat inactivated and filtered animal serum containing a detergent and a preservative.Used in ELISA assays for diluting standards, reagents and for blocking non-specific interactions. 1 * 100 mL
COMMANDE
Développer tout / Réduire tout
Heat inactivated and filtered animal serum containing a detergent and a preservative.Used in ELISA assays for diluting standards, reagents and for blocking non-specific interactions. 1 * 100 mL ORIGAR100016 1 * 100 mL
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou