Human Tagged ORF Clone Lentiviral Particle, ICOS
Fournisseur: OriGene
RC207031L3V
ORIGRC207031L3VEA 1120 EUR InStock
ORIGRC207031L3V
Human Tagged ORF Clone Lentiviral Particle, ICOS
Vecteurs, plasmides et librairies
Lenti ORF particles, ICOS (Myc-DDK tagged) - Human inducible T-cell co-stimulator (ICOS), 200 µl, >10^7 TU/ml.
The molecular sequence of this clone aligns with the gene accession number as a point of reference only. However, individual transcript sequences of the same gene can differ through naturally occurring variations (e.g. polymorphisms), each with its own valid existence. This clone is substantially in agreement with the reference, but a complete review of all prevailing variants is recommended prior to use.
This clone was engineered to express the complete ORF with an expression tag. Expression varies depending on the nature of the gene.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité