ORF Clones, GABA B receptor 1 (GABBR1), human tagged, TrueORF®
Fournisseur: OriGene
RC214114
ORIGRC214114EA 1690 EUR InStock
ORIGRC214114
ORF Clones, GABA B receptor 1 (GABBR1), human tagged, TrueORF®
Vecteurs, plasmides et librairies
GABA B receptor 1 (GABBR1), human tagged ORF clone was engineered to express the complete ORF with an expression tag. Expression varies depending on the nature of the gene.
This gene encodes a receptor for gamma-aminobutyric acid (GABA), which is the main inhibitory neurotransmitter in the mammalian central nervous system. This receptor functions as a heterodimer with GABA(B) receptor. Defects in this gene may underlie brain disorders such as schizophrenia and epilepsy. Alternative splicing generates multiple transcript variants, but the full-length nature of some of these variants has not been determined.
The ORF clone is ion-exchange column purified, transfection-ready dried plasmid DNA.
Information de livraison: Includes 2 vector sequencing primers.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité